Cobalt(II) meso-tetraphenylporphine ≥97%

Supplier: Ambeed
Warning

Synonyms: Cobalt(II) TPP, Cobalt(II) Tetraphenylporphyrin, 5,10,15,20-Tetraphenyl-21H,23H-porphine Cobalt(II)

A686346-1G A686346-250MG A686346-25G A686346-5G
77255-036EA 37 USD
77255-036 77255-038 77255-040 77255-042
Cobalt(II) meso-tetraphenylporphine ≥97%
Cobalt(II) meso-tetraphenylporphine

Precaución: for Research Use Only

Formula: C₄₄H₂₈CoN₄
MW: 671.65 g/mol
MDL Number: MFCD00010725
CAS Number: 14172-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR