Benzene-1,3,5-triyltriboronic acid 95%

Supplier: Ambeed
Warning

A686226-100MG A686226-1G A686226-250MG
77255-030EA 28.84 USD
77255-030 77255-032 77255-034
Benzene-1,3,5-triyltriboronic acid 95%
Benzene-1,3,5-triyltriboronic acid

Precaución: for Research Use Only

Formula: C₆H₉B₃O₆
MW: 209.56 g/mol
MDL Number: MFCD20275289
CAS Number: 89641-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR