3,3-Diphenyl-1-propanol 98%

Supplier: Ambeed
Warning

A686177-1G A686177-25G A686177-5G
77280-746EA 18 USD
77280-746 77280-748 77280-750
3,3-Diphenyl-1-propanol 98%
3,3-Diphenyl-1-propanol

Precaución: for Research Use Only

Formula: C₁₅H₁₆O
MDL Number: MFCD00002930
CAS Number: 20017-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR