3-Bromo-2,5-dichlorobenzoic acid 95%

Supplier: Ambeed
Warning

A685490-100MG A685490-1G A685490-250MG
77243-346EA 42.23 USD
77243-346 77243-348 77243-350
3-Bromo-2,5-dichlorobenzoic acid 95%
3-Bromo-2,5-dichlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrCl₂O₂
MW: 269.90 g/mol
MDL Number: MFCD07780678
CAS Number: 855202-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR