1-(Cyanoacetyl)piperidine 95%

Supplier: Ambeed
Warning

Synonyms: 3-Oxo-3-(piperidin-1-yl)propanenitrile

A685211-10G A685211-1G
77243-342EA 59.74 USD
77243-342 77243-344
1-(Cyanoacetyl)piperidine 95%
1-(Cyanoacetyl)piperidine

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O
MW: 152.20 g/mol
MDL Number: MFCD00006487
CAS Number: 15029-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR