5-(Bromomethyl)-2-chlorobenzonitrile 97%

Supplier: Ambeed
Danger

A684992-100MG A684992-250MG A684992-5G
77267-404EA 64 USD
77267-404 77267-406 77267-408
5-(Bromomethyl)-2-chlorobenzonitrile 97%
5-(Bromomethyl)-2-chlorobenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrClN
MDL Number: MFCD18392463
CAS Number: 99613-64-6
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR