2-Methyl-1-(naphthalen-2-yl)propan-2-amine 98+%

Supplier: Ambeed
Warning

A684902-100MG A684902-1G A684902-250MG A684902-5G
77267-396EA 29 USD
77267-396 77267-398 77267-400 77267-402
2-Methyl-1-(naphthalen-2-yl)propan-2-amine 98+%
2-Methyl-1-(naphthalen-2-yl)propan-2-amine

Precaución: for Research Use Only

Formula: C₁₄H₁₇N
MDL Number: MFCD11519113
CAS Number: 198226-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR