N-(3-Amino-4-methylphenyl)-3-(trifluoromethyl)benzamide 97%

Supplier: Ambeed
Warning

A684796-100MG A684796-1G A684796-250MG
77255-024EA 21 USD
77255-024 77255-026 77255-028
N-(3-Amino-4-methylphenyl)-3-(trifluoromethyl)benzamide 97%
N-(3-Amino-4-methylphenyl)-3-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₅H₁₃F₃N₂O
MDL Number: MFCD09930727
CAS Number: 30069-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR