N-(2-Methyl-5'-morpholino-6'-((tetrahydro-2H-pyran-4-yl)oxy)-[3,3'-bipyridin]-5-yl)-3-(trifluoromethyl)benzamide 97%
Supplier: Ambeed
|
Warning
|
A684624-10MG
A684624-1MG
A684624-25MG
A684624-50MG
A684624-5MG
77267-370EA
93
USD
77267-370
77267-372
77267-374
77267-376
77267-378
N-(2-Methyl-5'-morpholino-6'-((tetrahydro-2H-pyran-4-yl)oxy)-[3,3'-bipyridin]-5-yl)-3-(trifluoromethyl)benzamide 97%
N-(2-Methyl-5'-morpholino-6'-((tetrahydro-2H-pyran-4-yl)oxy)-[3,3'-bipyridin]-5-yl)-3-(trifluoromethyl)benzamide
Precaución: for Research Use Only
Formula:
C₂₈H₂₉F₃N₄O₄ |
MDL Number:
MFCD30534393 CAS Number: 1628838-42-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...