2,2,3,3-Tetrafluoro-1-propanol 98%

Supplier: Ambeed
Danger

Synonyms: 2,2,3,3-Tetrafluoropropan-1-ol, 1H,1H,3H-Tetrafluoro-1-propanol

A684449-100G A684449-10G A684449-25G
77255-018EA 10.3 USD
77255-018 77255-020 77255-022
2,2,3,3-Tetrafluoro-1-propanol 98%
2,2,3,3-Tetrafluoro-1-propanol

Precaución: for Research Use Only

Formula: C₃H₄F₄O
MW: 132.06 g/mol
MDL Number: MFCD00004676
CAS Number: 76-37-9
UN: 1986
ADR: 3,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR