N-((2S,3R,4R,5R,6R)-2,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)acetamide 97%

Supplier: Ambeed
Warning

A684383-100MG A684383-1G A684383-250MG
77255-012EA 5.15 USD
77255-012 77255-014 77255-016
N-((2S,3R,4R,5R,6R)-2,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)acetamide 97%
N-((2S,3R,4R,5R,6R)-2,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)acetamide

Precaución: for Research Use Only

Formula: C₈H₁₅NO₆
MW: 221.21 g/mol
MDL Number: MFCD00065372
CAS Number: 14215-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR