2,4-Difluoro-5-nitrobenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A684093-10G
A684093-1G
A684093-25G
A684093-5G
77255-004EA
28
USD
77255-004
77255-006
77255-008
77255-010
2,4-Difluoro-5-nitrobenzoic acid 97%
2,4-Difluoro-5-nitrobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₃F₂NO₄ |
MDL Number:
MFCD04115713 CAS Number: 153775-33-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...