(S)-2,2'-Bis[bis(3,5-trifluoromethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl 98%

Supplier: Ambeed
Warning

Synonyms: (S)-BTFM-Garphos, (S)-2,2'-Bis[bis(3,5-trifluoromethylphenyl)phosphino]-4,4',6,6'-tetramethoxy)-1,1'-biphenyl, (S)-(4,4',6,6'-Tetramethoxybiphenyl-2,2'-diyl)bis(bis[3,5-bis(trifluoromethyl)phenyl]phosphine)

A683337-100MG A683337-1G A683337-250MG
77254-994EA 115.36 USD
77254-994 77254-996 77254-998
(S)-2,2'-Bis[bis(3,5-trifluoromethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl 98%
(S)-2,2'-Bis[bis(3,5-trifluoromethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl

Precaución: for Research Use Only

Formula: C₄₈H₂₈F₂₄O₄P₂
MW: 1186.64 g/mol
MDL Number: MFCD19443625
CAS Number: 1365531-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR