6-Bromo-2,3-dihydro-1H-pyrrolo[3,2-b]pyridine 95%

Supplier: Ambeed
Warning

A682681-100MG A682681-250MG A682681-1G
77280-728EA 90.64 USD
77280-728 77280-730 77749-406
6-Bromo-2,3-dihydro-1H-pyrrolo[3,2-b]pyridine 95%
6-Bromo-2,3-dihydro-1H-pyrrolo[3,2-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₇BrN₂
MW: 199.04 g/mol
MDL Number: MFCD18250825
CAS Number: 1260663-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR