1-Methyl-1H-indazole-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A682646-1G A682646-100G A682646-5G A682646-25G
77280-726EA 17.51 USD
77280-726 77617-838 77617-840 77749-405
1-Methyl-1H-indazole-3-carboxylic acid 98%
1-Methyl-1H-indazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.17 g/mol
MDL Number: MFCD00272569
CAS Number: 50890-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR