Ethyl-3,4-diaminobenzoate 97%

Supplier: Ambeed
Warning

A681879-100G A681879-10G A681879-1G A681879-25G A681879-5G
77254-978EA 101 USD
77254-978 77254-980 77254-982 77254-984 77254-986
Ethyl-3,4-diaminobenzoate 97%
Ethyl-3,4-diaminobenzoate

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₂
MDL Number: MFCD00179337
CAS Number: 37466-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR