2-(Trifluoromethyl)quinoline 97%

Supplier: Ambeed
Warning

A680867-100MG A680867-1G A680867-250MG
77280-712EA 19.57 USD
77280-712 77280-714 77280-716
2-(Trifluoromethyl)quinoline 97%
2-(Trifluoromethyl)quinoline

Precaución: for Research Use Only

Formula: C₁₀H₆F₃N
MDL Number: MFCD01862002
CAS Number: 347-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR