Tirapazamine 98%

Supplier: Ambeed
Warning

Synonyms: Tirazamin, 3-Aminobenzo[e][1,2,4]triazine 1,4-Dioxide, 4-Hydroxy-1-oxido-1,2,4-benzotriazin-1-ium-3-imine,SR 259075,SR 4233,Tirazone,Win 59075, SR 259075

A680596-10MG A680596-1MG A680596-100MG A680596-250MG A680596-50MG
77280-708EA 46.35 USD
77280-708 77280-710 77749-401 77749-402 77749-403
Tirapazamine 98%
Tirapazamine

Precaución: for Research Use Only

Formula: C₇H₆N₄O₂
MW: 178.15 g/mol
MDL Number: MFCD00132954
CAS Number: 27314-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR