2-Fluorobenzophenone 98%

Supplier: Ambeed
Warning

Synonyms: o-Fluorobenzophenone

A680076-10G A680076-25G A680076-5G
77267-362EA 13.39 USD
77267-362 77267-364 77749-399
2-Fluorobenzophenone 98%
2-Fluorobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₉FO
MW: 200.21 g/mol
MDL Number: MFCD00000318
CAS Number: 342-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR