1-Ethyl-1H-pyrazole 98%

Supplier: Ambeed
Danger

Synonyms: 1-Ethylpyrazole

A679992-1G A679992-25G A679992-5G
77243-322EA 9 USD
77243-322 77243-324 77243-326
1-Ethyl-1H-pyrazole 98%
1-Ethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₅H₈N₂
MW: 96.1304 g/mol
MDL Number: MFCD03444367
CAS Number: 2817-71-2
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR