2,2,2-Trifluoro-N-phenylacetamide 97%

Supplier: Ambeed
Warning

A679635-10G A679635-1G A679635-5G
77254-972EA 125.66 USD
77254-972 77254-974 77254-976
2,2,2-Trifluoro-N-phenylacetamide 97%
2,2,2-Trifluoro-N-phenylacetamide

Precaución: for Research Use Only

Formula: C₈H₆F₃NO
MW: 189.1346 g/mol
MDL Number: MFCD00196027
CAS Number: 404-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR