3-[(4S)-2,2-Dimethyl-1,3-dioxolan-4-yl]propanol 95%

Supplier: Ambeed
Warning

Synonyms: (S)-3-(2,2-Dimethyl-1,3-dioxolan-4-yl)propan-1-ol

A679083-1G A679083-250MG
77280-692EA 530.45 USD
77280-692 77280-694
3-[(4S)-2,2-Dimethyl-1,3-dioxolan-4-yl]propanol 95%
3-[(4S)-2,2-Dimethyl-1,3-dioxolan-4-yl]propanol

Precaución: for Research Use Only

Formula: C₈H₁₆O₃
MW: 160.213 g/mol
MDL Number: MFCD01631673
CAS Number: 51268-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR