tert-Butyl (4-iodobutyl)carbamate 98%

Supplier: Ambeed
Warning

A678765-1G A678765-250MG
77254-970EA 32 USD
77254-970 77747-374
tert-Butyl (4-iodobutyl)carbamate 98%
tert-Butyl (4-iodobutyl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₈INO₂
MDL Number: MFCD09951820
CAS Number: 262278-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR