Morpholin-2-ylmethanol 95%

Supplier: Ambeed
Warning

A677872-100MG A677872-1G A677872-250MG
77267-332EA 64.89 USD
77267-332 77267-334 77267-336
Morpholin-2-ylmethanol 95%
Morpholin-2-ylmethanol

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂
MDL Number: MFCD03426271
CAS Number: 103003-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR