2-Chloro-1,4-difluorobenzene 98%

Supplier: Ambeed
Danger

Synonyms: 2,5-Difluorochlorobenzene

A677781-10G A677781-25G A677781-5G
77243-306EA 15 USD
77243-306 77243-308 77747-373
2-Chloro-1,4-difluorobenzene 98%
2-Chloro-1,4-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃ClF₂
MW: 148.54 g/mol
Boiling Pt: 128
Melting Pt: -24.6
MDL Number: MFCD00042571
CAS Number: 2367-91-1
UN: 1993
ADR: 3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR