N-(2-Methoxy-4-morpholinophenyl)-2-(pyridin-3-yl)thiazole-4-carboxamide 98%

Supplier: Ambeed
Warning

A677710-10MG A677710-1MG A677710-50MG A677710-5MG
77243-298EA 278.1 USD
77243-298 77243-300 77243-302 77243-304
N-(2-Methoxy-4-morpholinophenyl)-2-(pyridin-3-yl)thiazole-4-carboxamide 98%
N-(2-Methoxy-4-morpholinophenyl)-2-(pyridin-3-yl)thiazole-4-carboxamide

Precaución: for Research Use Only

Formula: C₂₀H₂₀N₄O₃S
MW: 396.46 g/mol
MDL Number: MFCD22124485
CAS Number: 1042672-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR