(2,4-Dioxo-3,4-dihydro-2H-pyrimidin-1-yl)acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-(2,4-Dioxo-3,4-dihydropyrimidin-1(2H)-yl)acetic acid

A677687-1G A677687-250MG A677687-5G
77243-292EA 52.53 USD
77243-292 77243-294 77243-296
(2,4-Dioxo-3,4-dihydro-2H-pyrimidin-1-yl)acetic acid 95%
(2,4-Dioxo-3,4-dihydro-2H-pyrimidin-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₆H₆N₂O₄
MW: 170.13 g/mol
MDL Number: MFCD00458007
CAS Number: 4113-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR