5-Acetamido-2-nitrophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (5-Acetamido-2-nitro)benzeneboronicacid

A677555-1G A677555-250MG A677555-5G
77254-956EA 85.49 USD
77254-956 77254-958 77254-960
5-Acetamido-2-nitrophenylboronic acid 98%
5-Acetamido-2-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BN₂O₅
MW: 223.9800 g/mol
MDL Number: MFCD06659829
CAS Number: 78887-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR