3,6-Dibromo-9-ethylcarbazole 98%

Supplier: Ambeed
Warning

A677148-1G A677148-250MG A677148-25G A677148-5G
77280-672EA 27 USD
77280-672 77280-674 77280-676 77280-678
3,6-Dibromo-9-ethylcarbazole 98%
3,6-Dibromo-9-ethylcarbazole

Precaución: for Research Use Only

Formula: C₁₄H₁₁Br₂N
MDL Number: MFCD00138134
CAS Number: 33255-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR