3-Amino-N,N-dimethylbenzamide 95%

Supplier: Ambeed
Warning

A677031-1G A677031-250MG A677031-5G
77280-666EA 53.56 USD
77280-666 77280-668 77280-670
3-Amino-N,N-dimethylbenzamide 95%
3-Amino-N,N-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O
MW: 164.2044 g/mol
MDL Number: MFCD01168905
CAS Number: 33322-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR