2,2-Bis(4-hydroxy-3,5-dimethylphenyl)propane 98%

Supplier: Ambeed
Warning

Synonyms: Tetramethylbisphenol A, 4,4'-(Propane-2,2-diyl)bis(2,6-dimethylphenol), 4,4'-Isopropylidenebis(2,6-dimethylphenol)

A676008-100G A676008-1G A676008-25G A676008-5G
77254-944EA 288.4 USD
77254-944 77254-946 77254-948 77254-950
2,2-Bis(4-hydroxy-3,5-dimethylphenyl)propane 98%
2,2-Bis(4-hydroxy-3,5-dimethylphenyl)propane

Precaución: for Research Use Only

Formula: C₁₉H₂₄O₂
MW: 284.40 g/mol
MDL Number: MFCD00068238
CAS Number: 5613-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR