4'-Bromo-3'-(trifluoromethyl)acetanilide 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-Bromo-3-(trifluoromethyl)phenyl)acetamide

A675731-1G A675731-250MG
77267-314EA 11.33 USD
77267-314 77267-316
4'-Bromo-3'-(trifluoromethyl)acetanilide 98%
4'-Bromo-3'-(trifluoromethyl)acetanilide

Precaución: for Research Use Only

Formula: C₉H₇BrF₃NO
MW: 282.07 g/mol
MDL Number: MFCD00052361
CAS Number: 41513-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR