Dibenzo[b,d]furan-1-amine 97%

Supplier: Ambeed
Warning

A675667-1G A675667-25G
77267-310EA 21.63 USD
77267-310 77267-312
Dibenzo[b,d]furan-1-amine 97%
Dibenzo[b,d]furan-1-amine

Precaución: for Research Use Only

Formula: C₁₂H₉NO
MW: 183.20 g/mol
MDL Number: MFCD01461772
CAS Number: 50548-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR