2-Hexyl-1-decanol 98%

Supplier: Ambeed
Warning

Synonyms: Guerbet hexadecanol, Hexyldecanol, 2-Hexyldecanol

A674560-100G A674560-25G A674560-500G
77267-296EA 22 USD
77267-296 77267-298 77267-300
2-Hexyl-1-decanol 98%
2-Hexyl-1-decanol

Precaución: for Research Use Only

Formula: C₁₆H₃₄O
MW: 242.45 g/mol
MDL Number: MFCD00060903
CAS Number: 2425-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR