4-Bromo-2-iodopyridine 97%

Supplier: Ambeed
Warning

A674250-100MG A674250-250MG A674250-1G
77293-126EA 78.28 USD
77293-126 77293-128 77552-054
4-Bromo-2-iodopyridine 97%
4-Bromo-2-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₃BrIN
MW: 283.89 g/mol
MDL Number: MFCD07368063
CAS Number: 100523-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR