(S)-(+)-2,2'-Bis[di(3,5-di-t-butyl-4-methoxyphenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl 98%

Supplier: Ambeed
Warning

A673908-100MG A673908-1G A673908-250MG A673908-50MG
77254-922EA 135.96 USD
77254-922 77254-924 77254-926 77254-928
(S)-(+)-2,2'-Bis[di(3,5-di-t-butyl-4-methoxyphenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl 98%
(S)-(+)-2,2'-Bis[di(3,5-di-t-butyl-4-methoxyphenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₇₄H₁₀₄O₆P₂
MW: 1151.5615 g/mol
MDL Number: MFCD09265029
CAS Number: 910134-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR