2-Ethoxypyrazine 99+%

Supplier: Ambeed
Warning

A673657-100G A673657-25G
77267-288EA 51 USD
77267-288 77267-290
2-Ethoxypyrazine 99+%
2-Ethoxypyrazine

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MDL Number: MFCD00014609
CAS Number: 38028-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR