3,4-Dibromo-1-methyl-2,5-dihydro-1H-pyrrole-2,5-dione 95%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dibromo-N-methylmaleimide

A673181-100MG A673181-250MG
77254-914EA 33.99 USD
77254-914 77254-916
3,4-Dibromo-1-methyl-2,5-dihydro-1H-pyrrole-2,5-dione 95%
3,4-Dibromo-1-methyl-2,5-dihydro-1H-pyrrole-2,5-dione

Precaución: for Research Use Only

Formula: C₅H₃Br₂NO₂
MW: 268.891 g/mol
MDL Number: MFCD00102284
CAS Number: 3005-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR