ω-(4-Chlorophenyl)acetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Chlorophenyl)acetophenone

A673139-100MG A673139-1G A673139-250MG
77243-258EA 13.39 USD
77243-258 77243-260 77243-262
ω-(4-Chlorophenyl)acetophenone 98%
ω-(4-Chlorophenyl)acetophenone

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClO
MW: 230.6895 g/mol
MDL Number: MFCD02260686
CAS Number: 6332-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR