2-Chloro-3-(trifluoromethyl)quinoxaline 97%
Supplier: Ambeed
|
Danger
|
A672780-250MG
A672780-5G
77267-280EA
29
USD
77267-280
77267-282
2-Chloro-3-(trifluoromethyl)quinoxaline 97%
2-Chloro-3-(trifluoromethyl)quinoxaline
Precaución: for Research Use Only
Formula:
C₉H₄ClF₃N₂ |
MDL Number:
MFCD00243891 CAS Number: 254732-51-9 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...