4'-Methyl-2,2':6',2''-terpyridine 98%

Supplier: Ambeed
Warning

A672642-100MG A672642-250MG A672642-1G
77243-254EA 57.68 USD
77243-254 77243-256 77573-168
4'-Methyl-2,2':6',2''-terpyridine 98%
4'-Methyl-2,2':6',2''-terpyridine

Precaución: for Research Use Only

Formula: C₁₆H₁₃N₃
MW: 247.29 g/mol
MDL Number: MFCD06796968
CAS Number: 72036-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR