1-Chloro-4-ethylbenzene 97%

Supplier: Ambeed
Danger

Synonyms: 4-Chloro(ethylbenzene)

A672242-10G A672242-5G
77243-250EA 37.08 USD
77243-250 77243-252
1-Chloro-4-ethylbenzene 97%
1-Chloro-4-ethylbenzene

Precaución: for Research Use Only

Formula: C₈H₉Cl
MW: 140.61 g/mol
MDL Number: MFCD00040855
CAS Number: 622-98-0
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR