1-(4-Amino-2-chloropyridin-3-yl)ethanone 97%

Supplier: Ambeed
Danger

A672104-100MG A672104-1G A672104-250MG A672104-5G
77243-242EA 106.09 USD
77243-242 77243-244 77243-246 77243-248
1-(4-Amino-2-chloropyridin-3-yl)ethanone 97%
1-(4-Amino-2-chloropyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O
MW: 170.59 g/mol
MDL Number: MFCD22547429
CAS Number: 1393573-67-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR