(1S,2S)-N,N-Bis[2-(di-p-tolylphosphino)benzyl]cyclohexane-1,2-diamine ≥98%, ee 99%

Supplier: Ambeed
Warning

A671565-100MG A671565-1G A671565-250MG A671565-5G
77254-906EA 31.93 USD
77254-906 77254-908 77254-910 77254-912
(1S,2S)-N,N-Bis[2-(di-p-tolylphosphino)benzyl]cyclohexane-1,2-diamine ≥98%, ee 99%
(1S,2S)-N,N-Bis[2-(di-p-tolylphosphino)benzyl]cyclohexane-1,2-diamine

Precaución: for Research Use Only

Formula: C₄₈H₅₂N₂P₂
MW: 718.88 g/mol
MDL Number: MFCD17014024
CAS Number: 1224727-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR