endo-Bicyclo[2.2.1]heptan-2-amine hydrochloride 95%

Supplier: Ambeed
Warning

A671323-1G A671323-250MG A671323-5G
77267-272EA 336 USD
77267-272 77267-274 77267-276
endo-Bicyclo[2.2.1]heptan-2-amine hydrochloride 95%
endo-Bicyclo[2.2.1]heptan-2-amine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄ClN
MDL Number: MFCD13193751
CAS Number: 65481-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR