2-Bromo-N-methyl-6-nitroaniline 98%

Supplier: Ambeed
Warning

A671301-1G A671301-250MG
77243-238EA 197.76 USD
77243-238 77243-240
2-Bromo-N-methyl-6-nitroaniline 98%
2-Bromo-N-methyl-6-nitroaniline

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₂
MW: 231.05 g/mol
MDL Number: MFCD00090077
CAS Number: 1004618-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR