5-(Methylthio)-1,3,4-thiadiazol-2-amine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-5-(methylthio)-1,3,4-thiadiazole
A670866-100G
A670866-10G
A670866-1G
A670866-25G
77254-898EA
741.6
USD
77254-898
77254-900
77254-902
77254-904
5-(Methylthio)-1,3,4-thiadiazol-2-amine 98%
5-(Methylthio)-1,3,4-thiadiazol-2-amine
Precaución: for Research Use Only
Formula:
C₃H₅N₃S₂ MW: 147.22 g/mol |
MDL Number:
MFCD00175752 CAS Number: 5319-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...