6-Chloro-2-(trifluoromethyl)-3H-imidazo[4,5-b]pyridine 97%

Supplier: Ambeed
Warning

A670826-100MG A670826-1G A670826-250MG
77243-232EA 56 USD
77243-232 77243-234 77243-236
6-Chloro-2-(trifluoromethyl)-3H-imidazo[4,5-b]pyridine 97%
6-Chloro-2-(trifluoromethyl)-3H-imidazo[4,5-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₃ClF₃N₃
MDL Number: MFCD00223328
CAS Number: 13577-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR