(E)-3-(4-(Trifluoromethoxy)phenyl)acrylic acid 95%

Supplier: Ambeed
Warning

A670229-25G A670229-5G
77267-268EA 361.53 USD
77267-268 77267-270
(E)-3-(4-(Trifluoromethoxy)phenyl)acrylic acid 95%
(E)-3-(4-(Trifluoromethoxy)phenyl)acrylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇F₃O₃
MW: 232.15 g/mol
MDL Number: MFCD00066338
CAS Number: 199679-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR