4-Chloro-6-nitroquinoline-3-carbonitrile 97%

Supplier: Ambeed
Warning

A670130-100MG A670130-250MG
77293-104EA 75.19 USD
77293-104 77293-106
4-Chloro-6-nitroquinoline-3-carbonitrile 97%
4-Chloro-6-nitroquinoline-3-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₄ClN₃O₂
MW: 233.61 g/mol
MDL Number: MFCD09261363
CAS Number: 214470-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR